Learn More
Ginkgolide A, MedChemExpress
MedChemExpress Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.
52.65 EUR - 172.80 EUR
Specifications
Chemical Name or Material | Ginkgolide A |
---|---|
CAS | 15291-75-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C20H24O9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16454337
|
Medchem Express
HY-B0355 |
50 mg |
172.80 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16418458
|
Medchem Express
HY-B0355 |
10 mg |
52.65 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.Specifications
Ginkgolide A | |
White | |
C20H24O9 | |
DMSO : 100 mg/mL (244.86 mM; Need ultrasonic) | |
408.4 | |
98.0% | |
Solid |
15291-75-5 | |
Research | |
BN-52020 | |
O=C1[C@@H](C)[C@]2(O)[C@@]3(C4=O)C(C5([C@@H](O)C(O6)=O)C6O3)([C@H](O4)C[C@H]5C(C)(C)C)C[C@]2([H])O1 | |
408.4 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.