missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Assistent™ Glass Instrument Trays
Sterilizable in hot air up to +180°C. Karl Hecht™ Assistent™ Glass Instrument Trays are excellent for cleaning, sterilizing and storing laboratory instruments.
Brand: Karl Hecht™ 42900003
This item is not returnable.
View return policy
Description
- Autoclavable
- Available in a range of sizes
- Available with or without stainless steel cover
Specifications
200 x 100 x 50 mm | |
200 mm | |
Tray | |
Without cover |
50 mm | |
Glass | |
100 mm | |
Resistant to hot-air sterilization up to +180°C / +356°F |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Assistent™ Glass Instrument Trays > Dimensions (L x W x H): 200 x 100 x 50mm; Product Type: No cover
Spot an opportunity for improvement?Share a Content Correction