missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Glass Instrument Trays

Sterilizable in hot air up to +180°C. Karl Hecht™ Assistent™ Glass Instrument Trays are excellent for cleaning, sterilizing and storing laboratory instruments.

Brand:  Karl Hecht™ 42910003

 View more versions of this product

Product Code. 10749691

  • 72.00 EUR / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable
  • Available in a range of sizes
  • Available with or without stainless steel cover
Specifications

Specifications

200 x 100 x 50 mm
200 mm
Tray
With cover
Rectangular
50 mm
Glass, Stainless Steel
100 mm
Resistant to hot-air sterilization up to +180°C / +356°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Hecht™ Assistent™ Glass Instrument Trays > Dimensions (L x W x H): 200 x 100 x 50mm; Product Type: With cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.