Learn More
Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
Brand:  Fisherbrand™ FB51750
 View more versions of this product
Product Code. 11992128
Description
Description
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
  • Easy-open stopper helps to reduce pressure buildup during processing.
  • Constructed in chemical resistant borosilicate glass.
  • ISO 4800 Certified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
ISO 4800
Glass
19/26
Borosilicate Glass
1PK
Glass, Plastic
Separating funnel
Documents
One moment while we fetch your results.
Product Title
Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock > Capacity: 50mL