Learn More
Welch™ Glass Vaccum Fluivac Station
Brand:  Welch™ 112039
Product Code. 11887323
Specifications
1 Each
Vaccum Fluivac Station
Glass
5 L
Documents
One moment while we fetch your results.
Certificates
View accessory products
Product Title
Welch™ Glass Vaccum Fluivac Station > Flow Rate: 63 l/min.