Learn More
PPAR alpha/NR1C1 Antibody - N-terminal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NB600-636
This item is not returnable.
View return policy
Description
PPAR alpha/NR1C1 Polyclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
PPAR alpha/NR1C1 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 | |
Rabbit | |
52.35 kDa | |
0.1 mg | |
Cancer, mTOR Pathway | |
5465 | |
Human, Mouse, Rat, Canine, Hamster, Hamster | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
1 mg/ml | |
Western Blot 1:500-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:75000-1:125000, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
P23204 | |
PPARA | |
PPAR alpha/NR1C1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-Terminal region near amino acids 1-25 of mouse PPAR alpha/NR1C1. (Uniprot: P23204) | |
Affinity Purified | |
RUO | |
Primary | |
No reactivity is expected against other subtypes of PPAR. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
PPAR alpha/NR1C1 Antibody - N-terminal, Novus Biologicals™ > 0.1mg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction