missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Organic compounds
- Unclassified Organic Compounds
- Glutathione Reduced Crystalline MP Biomedicals
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Glutathione Reduced Crystalline MP Biomedicals
Product Code. 11314269 Shop All MP Biomedicals Products100 g
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
This item is not returnable.
View return policy
Product Code. 11314269
Brand: MP Biomedicals 0210181480
This item is not returnable.
View return policy
Specifications
Chemical Identifiers
CAS | 70-18-8 |
Molecular Formula | C10H17N3O6S |
Molecular Weight (g/mol) | 307.321 |
MDL Number | MFCD00065939 |
InChI Key | RWSXRVCMGQZWBV-WDSKDSINSA-N |
Synonym | glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion |
PubChem CID | 124886 |
ChEBI | CHEBI:16856 |
IUPAC Name | (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid |
SMILES | C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N |
Specifications
Color | White |
Assay Percent Range | >98% |
Quantity | 100 g |
Formula Weight | 307.3 |
Physical Form | Powder |
Chemical Name or Material | Glutathione, Reduced |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10628343
-
Product Code. 11975721
-
-
Product Code. 11465202
-
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 3
-
Catalog No. 11482874
45,15 € / 2.50 L -
Catalog No. 11480673
Silicone oil, for oil baths, usable range from -40 to +200 Silicone oil, for oil baths, usable range from -40 to +200
286,38 € / 2500g -
Catalog No. 10687282
67,03 € / 1 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glutathione Reduced Crystalline MP Biomedicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction