Learn More
GNE-272, MedChemExpress
MedChemExpress GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.
Brand: Medchem Express HY-100726
Description
GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.Specifications
GNE-272 | |
White | |
C22H25FN6O2 | |
DMSO : 100 mg/mL (235.59 mM; Need ultrasonic) | |
424.47 | |
99.74% | |
Solid |
1936428-93-1 | |
Research | |
100 mg | |
CC(N1CCC2=C(C(NC3=C(F)C=C(C4=CN(C)N=C4)C=C3)=NN2[C@@H]5COCC5)C1)=O | |
424.47 | |
Research | |
Cancer-Kinase/protease |