Learn More
GNE-272, MedChemExpress
MedChemExpress GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.
Brand:  Medchem Express HY-100726
Encompass_Preferred
 View more versions of this product
Product Code. 16464426
  • 2511.00 EUR / 100mg
Description
GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.
Specifications
Specifications
GNE-272
White
C22H25FN6O2
DMSO : 100 mg/mL (235.59 mM; Need ultrasonic)
424.47
99.74%
Solid
1936428-93-1
Research
100 mg
CC(N1CCC2=C(C(NC3=C(F)C=C(C4=CN(C)N=C4)C=C3)=NN2[C@@H]5COCC5)C1)=O
424.47
Research
Cancer-Kinase/protease
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
GNE-272, MedChemExpress