Learn More
GRA Ex-25, MedChemExpress
MedChemExpress GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.

POA - 1324.35 EUR
Specifications
Chemical Name or Material | GRA Ex-25 |
---|---|
CAS | 307983-31-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C29H36F3N3O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490346
![]() |
Medchem Express
HY-50675 |
2 mg |
POA
2mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16488078
![]() |
Medchem Express
HY-50675 |
10 mg |
294.30 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16487846
![]() |
Medchem Express
HY-50675 |
100 mg |
1324.35 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16473997
![]() |
Medchem Express
HY-50675 |
50 mg |
882.90 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430548
![]() |
Medchem Express
HY-50675 |
5 mg |
194.40 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.Specifications
GRA Ex-25 | |
White | |
C29H36F3N3O5 | |
O=C(O)CCNC(C1=CC=C(CN([C@H]2CC[C@H](C(C)(C)C)CC2)C(NC3=CC=C(OC(F)(F)F)C=C3)=O)C=C1)=O | |
563.61 | |
Research | |
Metabolism-sugar/lipid metabolism |
307983-31-9 | |
Research | |
DMSO : ≥ 32 mg/mL (56.78 mM) | |
563.61 | |
98.09% | |
Solid |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.