Learn More
Thermo Scientific Chemicals Grade 25 Sapphire Spheres, Dia.: 5mm
Brand: Thermo Scientific Chemicals 042058.KD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sapphire Sphere | |
5 mm | |
Grade 25 | |
50 Pieces |