Learn More
GSK0660, MedChemExpress
MedChemExpress GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Brand:  Medchem Express HY-12377
Encompass_Preferred
 View more versions of this product
Product Code. 16475846
  • 1227.15 EUR / 100mg
Explore more special offers
Description
Description
GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Specifications
Specifications
GSK0660
Light Yellow
C19H18N2O5S2
DMSO : ≥ 49 mg/mL (117.09 mM)
418.49
99.55%
Solid
1014691-61-2
Research
100 mg
O=C(C1=C(S(=O)(NC2=CC=C(NC3=CC=CC=C3)C=C2OC)=O)C=CS1)OC
418.49
Research
Metabolism-protein/nucleotide metabolism