missing translation for 'onlineSavingsMsg'
Learn More

GSK0660, MedChemExpress

MedChemExpress GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.

105.30 EUR - 1227.15 EUR

Specifications

Chemical Name or Material GSK0660
CAS 1014691-61-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H18N2O5S2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475846
Encompass_Preferred
View Documents
Medchem Express
HY-12377
100 mg
1227.15 EUR
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16467847
Encompass_Preferred
View Documents
Medchem Express
HY-12377
5 mg
105.30 EUR
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16445128
Encompass_Preferred
View Documents
Medchem Express
HY-12377
10 mg
174.15 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16411367
Encompass_Preferred
View Documents
Medchem Express
HY-12377
50 mg
684.45 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Specifications

Specifications

GSK0660
Light Yellow
C19H18N2O5S2
O=C(C1=C(S(=O)(NC2=CC=C(NC3=CC=CC=C3)C=C2OC)=O)C=CS1)OC
418.49
Research
Metabolism-protein/nucleotide metabolism
1014691-61-2
Research
DMSO : ≥ 49 mg/mL (117.09 mM)
418.49
99.55%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.