Learn More
Checkers Guard Dog™ Low Profile
Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market. These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
278.00 EUR - 876.00 EUR
Specifications
Color Black, Orange
Width (Metric) 91 cm
View More Specs
Products 5
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17722418
View Documents
Checkers
PGDS0015OB
27 cm
278.00 EUR
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
17792408
View Documents
Checkers
PGDS0009OB
43 cm
427.00 EUR
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
17732408
View Documents
Checkers
PGDS0001OB
72 cm
876.00 EUR
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
17752408
View Documents
Checkers
PGDS0004OB
77 cm
383.00 EUR
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
17782408
View Documents
Checkers
PGDS0008OB
84 cm
411.00 EUR
Each
Estimated Shipment: 07-04-2025
Description
  • Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market
  • These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
Specifications
Specifications
Black, Orange
91 cm
Documents
Product Certifications
Product Content Correction
Product Title
Checkers Guard Dog™ Low Profile