Learn More
GW3965 hydrochloride, MedChemExpress
Product Code. 16415056
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16415056
Brand: Medchem Express HY10627A
MedChemExpress GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Specifications
Chemical Name or Material GW3965 hydrochloride
CAS 405911-17-3
Color White
Purity Grade Notes Research
Molecular Formula C33H32Cl2F3NO3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl
Molecular Weight (g/mol) 618.51
Formula Weight 618.51
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation