missing translation for 'onlineSavingsMsg'
Learn More

GW791343 dihydrochloride, MedChemExpress

MedChemExpress GW791343 dihydrochloride is a P2X7 allosteric modulator; exhibits species-specific activity and acts as a negative allosteric modulator of human P2X7 (pIC50 = 6.9 - 7.2).

Encompass_Preferred

117.45 EUR - 635.85 EUR

Specifications

Chemical Name or Material GW791343 dihydrochloride
CAS 1019779-04-4
Color White
Purity Grade Notes Research
Molecular Formula C20H26Cl2F2N4O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16442647
Encompass_Preferred
View Documents
Medchem Express
HY-15469
50 mg
635.85 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16419137
Encompass_Preferred
View Documents
Medchem Express
HY-15469
5 mg
117.45 EUR
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16416558
Encompass_Preferred
View Documents
Medchem Express
HY-15469
10 mg
201.15 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GW791343 dihydrochloride is a P2X7 allosteric modulator
Specifications

Specifications

GW791343 dihydrochloride
White
C20H26Cl2F2N4O
O=C(NC1=CC(CN2CCNCC2)=CC=C1C)CNC3=CC=C(F)C(F)=C3.[H]Cl.[H]Cl
447.35
Research
Neuroscience-Neuromodulation
1019779-04-4
Research
H2O : 100 mg/mL (223.54 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (44.71 mM; Need ultrasonic)
447.35
98.03%
Solid
Videos
Safety and Handling

Safety and Handling

H315∣H319∣H335

missing translation for 'shelfLife' : 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)

Recommended Storage : 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
GW791343 dihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.