Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
Brand: Thermo Scientific™ 75003308
806.77 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
400 mL Bottle/Centrilab Adapter | |
Swinging Bucket Rotor | |
4297 | |
Yes | |
Set of 2 |
Molded Polyamide Nylone | |
1 x 400 mL Bottle or Centrilab Each Adapter | |
Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series | |
Autoclavable |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
- Multiple Manufacturers