Learn More
Hamilton™ Microlab™ 500 Diluter/Dispenser System Accessories
Tubing Assembly, PTFE, FEP
47.80 EUR - 48.60 EUR
Specifications
Material | FEP, PTFE |
---|---|
Overall Length (English) | 35.43 in. |
Overall Length (Metric) | 90 cm |
For Use With | Microlab 500 Diluter/Dispenser |
Product Type | Tubing Assembly |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10278141
|
Hamilton™
240360 |
Tubing Assembly |
48.60 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11583154
|
Hamilton™
240130 |
Tubing Assembly |
47.80 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use with Hamilton Microlab 500 Diluter/Dispenser
Specifications
FEP, PTFE | |
90 cm | |
Tubing Assembly |
35.43 in. | |
Microlab 500 Diluter/Dispenser |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hamilton™ Microlab™ 500 Diluter/Dispenser System Accessories