Learn More
Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
BIOPHARMAGREENER_CHOICE
Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.
Brand:  Keystone Cleanroom Products COVMGTVKUB24X36
 View more versions of this product
Product Code. 17788357
  • 697.00 EUR / Pack of 50
Estimated Shipment: 26-06-2025
Description
  • All Tyvek construction for strength and microbial barrier
  • Designed for steam sterilization
  • Strong Factory seals
  • Heat Seal Closure
  • Recommended for sterilization of pharmaceutical equipment and components
  • Manufactured and packaged in a ISO 6 cleanroom
  • Improves the efficiency of the Parts Preparation Department
Specifications
Specifications
Autoclavable
None
Single-use
Tyvek
Yes
36 in.
24 in.
Autoclave Bag
COC
White
Pharmaceutical Equipment
Non-sterile
61 x 91.4 cm (24 x 36 in.)
91.4 cm
61 cm
Product Title
Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot > 24 x 36 in.