Learn More
Heidolph™ Hei-FLOW Gold Package
This package includes Hei-FLOW Advantage 01, pump head SP quick 1.6, 1 m each Tygon and silicone tubing.
Brand: Heidolph™ 523-51019-00
2806.96 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
This package includes:
- Hei-FLOW Advantage 01
- Pump head SP quick 1.6
- 1 m each Tygon™ (standard) and Silicone tubing (inner Ø 0.8 mm)
Specifications
EU Plug | |
Digital Variable Speed | |
LED | |
IP33 |
Special Offers
- Multiple Manufacturers
Product Content Correction
Product Title
Heidolph™ Hei-FLOW Gold Package > EU Plug
Spot an opportunity for improvement?Share a Content Correction