Learn More
Heidolph™ Instruments Hei-VAP Expert HL G6, EU-plug
The Hei-VAP Expert HL G6 allows for precise setting of rotation speed and heating bath temperature on the digital touch display or directly via the knobs on the operating panel.
Brand: Heidolph™ Instruments 573-01600-00
This item is not returnable.
View return policy
Description
Precise setting of rotation speed and heating bath temperature on the digital touch display or – for quick access during an active process – directly via the knobs on the operating panel.- Central operating panel with 5 in. touch display
- Hand lift model with G6 vertical glassware for reflux distillation
- Clear display of all values (Basic and Advanced modes)
- With timer, limitation of max. temperature, multilingual menu and text display for error messages
- Separate knobs for rotation and heating with locking function to prevent accidental readjusting
- LED ring lights and pictogram to display the activity even from a distance
- Integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller
- USB and Micro SD interfaces
- Optional display of vapor temperature
- Subsequent upgrade to Hei-VAP Expert Control possible, including vacuum and chiller control
Specifications
5 L | |
Hand Lift Rotary Evaporator | |
EU-plug |
Product Content Correction
Product Title
Heidolph™ Instruments Hei-VAP Expert HL G6, EU-plug > Type: Hand Lift Rotary Evaporator; Capacity (Metric): 5 L
Spot an opportunity for improvement?Share a Content Correction