missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hh-Ag1.5

Potent and high affinity Smo receptor agonist

229.00 EUR - 713.00 EUR

Specifications

Chemical Name or Material 3-Chloro-4,7-difluoro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide
CAS 612542-14-0
Target Smoothened
Molecular Formula C28H26ClF2N3OS
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18779976
View Documents
Tocris Bioscience™
7807/1
1 mg
229.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
18758716
View Documents Promotion Details
Tocris Bioscience™
7807/5
5 mg
713.00 EUR
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hh-Ag1.5 is a potent and high affinity Hedgehog (Hh) receptor agonist (EC50 = 1 nM; Ki values are 0.5 and 2.3 nM respectively, in a [3H]SAG-1.3 and [3H]Cyclopamine binding assay). It is used in protocols for the chemical reprogramming of mouse embryonic fibroblasts into neural stem cells. Also induces differentiation of hiPSCs into skin-derived precursor cells, spinal motor neurons and spinal sensory neurons.
Specifications

Specifications

3-Chloro-4,7-difluoro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide
612542-14-0
Smoothened
C28H26ClF2N3OS
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documents

Documents

Product Certifications