Learn More
Sartorius™ High Performance Glass Shield Secura26
High performance glass shield for use with laboratory balance.
Brand: Sartorius™ YHK01SQP
880.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used for increasing weighing performance of balances with readability of 0.002 mg
- (Supplied without a balance)
Specifications
Secura Laboratory Balances | |
Draft Shield |
Product Content Correction
Product Title
Sartorius™ High Performance Glass Shield Secura26 > High Performance Glass Shield Secura26; For Use With (Equipment): Secura Laboratory Balances; Type: Draft Shield
Spot an opportunity for improvement?Share a Content Correction