Learn More
MP Biomedicals High Unsaturated Fat Diet, Powder
High in safflower oil. High in unsaturated fats. 4.39 kcal/gm.
Brand:  MP Biomedicals 0296024310
 View more versions of this product
Product Code. 11437131
  • 648.72 EUR / 10kg
Description
  • High in Safflower oil
  • High in unsaturated fats
  • 4.39 kcal/gm
  • Contains casein, methionine, corn starch, mineral and vitamin fortification mix
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
10 kg
Refrigerated
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.