Learn More
Cytiva HiScale™ Column Tubes
Brand:  Cytiva 28-9666-52
 View more versions of this product
Product Code. 12425690
  • 638.00 EUR / Each
Estimated Shipment: 13-06-2025
Specifications
16 mm
Borosilicate Glass, Polycarbonate, Polyamide (Reinforced Glass Fiber), Ethylene Propylene Diene Poly
Documents
One moment while we fetch your results.
Product Title
Cytiva HiScale™ Column Tubes > I.D.: 16mm