missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific™

35.00 EUR - 147.00 EUR

Chemical Identifiers

CAS 38245-34-0
Molecular Formula C3Ho2O9
Molecular Weight (g/mol) 509.89
MDL Number MFCD00011497
InChI Key FZKKGPOEHOOXQE-UHFFFAOYSA-H
Synonym holmium carbonate hydrate, holmium iii carbonate hydrate, acmc-20akrc, holmium carbonate-water 2/3/1, holmium iii carbonate hydrate, reacton
PubChem CID 57350480
IUPAC Name holmium(3+);tricarbonate;hydrate
SMILES [Ho+3].[Ho+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342538
SDS View Documents
Thermo Scientific Alfa Aesar
014589.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11352538
SDS View Documents
Thermo Scientific Alfa Aesar
014589.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 227-584-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Holmium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.