Learn More
Sperian™ Honeywell™ LG Safety Goggles
Click to view available options
Clear Uncoated Polycarbonate Lens
Description
- Recommended for manufacturing and pharmaceutical work
- Lenses are colorless and anti-fog
- With 4 indirect vents and a wide field of vision
Specifications
Specifications
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Safety Spectacles |
For Use With (Equipment) | Industry Automotive and Part Manufacturer Building and Construction Food Industries Industrial Cleaning Laboratory Maintenance |
For Use With (Application) | Industry Automotive and Part Manufacturer Building and Construction Food Industries Industrial Cleaning Laboratory Maintenance |
Frame Color | Clear |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
Sperian™ Honeywell™ LG Safety Goggles >
Spot an opportunity for improvement?Share a Content Correction