missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hormones: b-Estradiol
Brand: MP Biomedicals™ 0219456501
This item is not returnable.
View return policy
Description
For research use only. Not intended for therapeutic use.

Specifications
15° to 30°C | |
Beta-Estradiol | |
1000 mg |
Crystalline powder | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- beta-Estradiol
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Reproductive toxicity Category 1A
Hazard Statement
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H362-May cause harm to breast-fed children.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : estradiol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Hormones: b-Estradiol > B-estradiol 1g
Spot an opportunity for improvement?Share a Content Correction