missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586155
This item is not returnable.
View return policy
Description
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
HSP40 | |
Polyclonal | |
Unconjugated | |
DNAJB1 | |
0610007I11Rik, DjB1, DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ heat shock protein family (Hsp40) member B1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, Dnajb1, HDJ1, hDj-1, Heat shock 40 kDa protein 1, Heat shock protein, HSP, HSP 40, HSP40, HSPF1, human DnaJ protein 1, NDAJB1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3337, 361384, 81489 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.2 | |
P25685, Q9QYJ3 | |
DNAJB1 | |
Synthetic peptide, corresponding to amino acids 270-320 of Human HSP40. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSP40 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction