missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ACE2 (aa1-615) Fc Chimera Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ RP87691
This item is not returnable.
View return policy
Description
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.Specifications
Q9BYF1 | |
Control, ELISA, Western Blot | |
59272 | |
Human ACE2 (aa1-615) Fc Chimera | |
HEK293 cells | |
RUO | |
ACE2 | |
Human | |
Human | |
Fc-Tag | |
>95% |
1 mg/mL | |
PBS with no preservative | |
110 kDa | |
50 μg | |
-20°Cor to +80°C if preferred | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
Protein | |
Unconjugated | |
Recombinant | |
Lyophilized |