Learn More
Invitrogen™ Human C6orf1 Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP95027
Description
Highest antigen sequence indentity to the following orthologs: Mouse (26%), Rat (26%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58046 (PA5-58046. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.
Specifications
Q86T20 | |
Blocking Assay, Control | |
221491 | |
100 μL | |
C6orf1; chromosome 6 open reading frame 1; LBH; Protein LBH; Small integral membrane protein 29; SMIM29; uncharacterized protein C6orf1 | |
SMIM29 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human C6orf1 Control Fragment | |
RUO | |
C6orf1 | |
Unconjugated | |
Recombinant | |
LRNCMRLSRSCSLTWETPRWYMAGRVATSTSGCHCWMSRRDLTPLPHPSEPGVLDCLGPCHLLPLLSP | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.