missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-10, Animal-Free Recombinant Protein, PeproTech®

Recombinant Protein

230.00 EUR - 3410.00 EUR

Specifications

Accession Number O15520
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2255
Molecular Weight (g/mol) 19.3 kDa
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17893413
View Documents Promotion Details
Gibco™
AF-100-26-100UG
100 μg
708.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
17893693
View Documents Promotion Details
Gibco™
AF-100-26-1MG
1 mg
3410.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
17863713
View Documents Promotion Details
Gibco™
AF-100-26-25UG
25 μg
230.00 EUR
25µg
Please to purchase this item. Need a web account? Register with us today!
17803703
View Documents
Gibco™
AF-100-26-500UG
500 μg
2040.00 EUR
500µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant Human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Human FGF-10, Animal-Free Recombinant Protein, PeproTech®

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.