Learn More
Gibco™ Human IL-3 Recombinant Protein, PeproTech®
Recombinant Protein
Brand: Gibco™ 200-03-250UG
725.36 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Recombinant Human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications
P08700 | |
protein with no preservative | |
15 kDa | |
250 μg | |
RUO | |
BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF | |
IL3 | |
Unconjugated | |
APMTQTTSLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
3562 | |
Human IL-3 | |
E. coli | |
<1 EU/ μg | |
IL-3 | |
The ED50 as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg | |
Recombinant | |
-20°C | |
Lyophilized |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.