missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7 Recombinant Protein, PeproTech®

Recombinant Protein

83.75 EUR - 5380.00 EUR

Specifications

For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Name Human IL-7
Source E. coli
Regulatory Status RUO
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17851383
View Documents Promotion Details
Gibco™
200-07-100UG
100 μg
1078.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
17810823
View Documents
Gibco™
200-07-10UG
10 μg
203.00 EUR
10µg
Please to purchase this item. Need a web account? Register with us today!
30089743
View Documents Promotion Details
Gibco™
200-07-01M
2 x 500 μg
5380.00 EUR
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
17821783
View Documents
Gibco™
200-07-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17881673
View Documents Promotion Details
Gibco™
200-07-250UG
250 μg
1882.00 EUR
250µg
Please to purchase this item. Need a web account? Register with us today!
17860153
View Documents
Gibco™
200-07-2UG
2 μg
83.75 EUR
2µg
Please to purchase this item. Need a web account? Register with us today!
17851073
View Documents Promotion Details
Gibco™
200-07-500UG
500 μg
3225.00 EUR
500µg
Please to purchase this item. Need a web account? Register with us today!
17800423
View Documents Promotion Details
Gibco™
200-07-50UG
50 μg
648.00 EUR
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

200-07-1MG will be provided as 2 x 500 μg (200-07-500UG). Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications

Specifications

ELISA, Functional Assay, Western Blot
Human IL-7
RUO
Il7
Recombinant
Lyophilized
protein with no preservative
E. coli
A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Unconjugated
MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Human IL-7 Recombinant Protein, PeproTech®

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.