Learn More
Invitrogen™ Human MIA-2 ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand: Invitrogen™ 900-K357K
This item is not returnable.
View return policy
Description
CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
Specifications
Q96PC5 | |
Lot-specific | |
ELISA | |
4253 | |
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 45 mins |
Lot-specific | |
HRP | |
Colorimetric Microplate Reader | |
Mia2 | |
HRP | |
RUO | |
CTAGE5 | |
24 hrs 30 mins |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Invitrogen™ Human MIA-2 ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP
Spot an opportunity for improvement?Share a Content Correction