Learn More
Invitrogen™ Human MIA-2 ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand:  Invitrogen™ 900-K357K
Product Code. 30087916
  • 382.00 EUR / Pack of 10
Description
CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q96PC5
Lot-specific
ELISA
4253
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
HRP
Colorimetric Microplate Reader
Mia2
HRP
RUO
CTAGE5
24 hrs 30 mins
Product Title
Invitrogen™ Human MIA-2 ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP