missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TPO (Thrombopoietin) Recombinant Protein, PeproTech®

Recombinant Protein

83.75 EUR - 4445.00 EUR

Specifications

Accession Number P40225
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 7066
Molecular Weight (g/mol) 18.6 kDa
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17859962
View Documents
Gibco™
300-18-100UG
100 μg
886.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
17811113
View Documents Promotion Details
Gibco™
300-18-10UG
10 μg
203.00 EUR
10µg
Please to purchase this item. Need a web account? Register with us today!
17830433
View Documents Promotion Details
Gibco™
300-18-1MG
1 mg
4445.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
17881713
View Documents Promotion Details
Gibco™
300-18-250UG
250 μg
1556.00 EUR
250µg
Please to purchase this item. Need a web account? Register with us today!
17850343
View Documents
Gibco™
300-18-2UG
2 μg
83.75 EUR
2µg
Please to purchase this item. Need a web account? Register with us today!
17871713
View Documents Promotion Details
Gibco™
300-18-500UG
500 μg
2665.00 EUR
500µg
Please to purchase this item. Need a web account? Register with us today!
17811813
View Documents Promotion Details
Gibco™
300-18-50UG
50 μg
534.00 EUR
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant Human TPO is a fully biologically active 174 amino acid polypeptide (18.6 kDa), which contains the erythropoietin-like domain of the full length TPO protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Human TPO (Thrombopoietin) Recombinant Protein, PeproTech®

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.