Learn More
Invitrogen™ Human ZNF717 Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP96472
Description
Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60742 (PA5-60742. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications
Q9BY31 | |
Blocking Assay, Control | |
100131827 | |
100 μL | |
krueppel-like factor X17; kruppel-like zinc finger factor X17; X17; zinc finger protein 717; zinc finger protein 838; ZNF717; ZNF838 | |
ZNF717 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human ZNF717 Control Fragment | |
RUO | |
ZNF717 | |
Unconjugated | |
Recombinant | |
PNLRLSGGSKKQVFSGICHRSLVELQEV | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.