missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human IL-28B Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
86.00 EUR - 2087.00 EUR
Specifications
Gene ID (Entrez) | 282617 |
---|---|
Molecular Weight (g/mol) | 24 kDa, monomer, non-glycosylated |
Species | Human |
Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16841386
|
Proteintech™
HZ-1245-10UG |
10 μg |
86.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16831386
|
Proteintech™
HZ-1245-100UG |
100 μg |
417.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16821386
|
Proteintech™
HZ-1245-1000UG |
1000 μg |
2087.00 EUR
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
282617 | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |
24 kDa, monomer, non-glycosylated | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically ≤ 1 ng/mL EC50 | |
Lyophilized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech™ HumanKine™ Recombinant Human IL-28B Protein