missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human IL-9 Protein

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

137.00 EUR - 3608.00 EUR

Specifications

Gene ID (Entrez) 3578
Molecular Weight (g/mol) 38 to 48 kDa, monomer, glycosylated
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16811386
View Documents
Proteintech™
HZ-1240-10UG
10 μg
137.00 EUR
10µg
Please to purchase this item. Need a web account? Register with us today!
16801386
View Documents
Proteintech™
HZ-1240-100UG
100 μg
722.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
16891376
View Documents
Proteintech™
HZ-1240-1000UG
1000 μg
3608.00 EUR
1000µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications

Specifications

3578
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%
38 to 48 kDa, monomer, glycosylated
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 1 ng/mL EC50
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Proteintech™ HumanKine™ Recombinant Human IL-9 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.