Learn More
Cytiva HyClone™ Porcine Serum (New Zealand)
Used to enhance cell culture performance, cost-efficient alternatives to Fetal Bovine Serum (FBS)
Brand: Cytiva SH30908.04
This item is not returnable.
View return policy
Description
Cytiva's porcine serum is sourced from New Zealand, filtered in the U.S. through multiple 200 nm pore size-rated filters and is gamma irradiated at 25 to 40 kGy after filtration.
Specifications
New Zealand | |
1 L | |
Irradiated | |
No | |
HyClone™ |
Porcine Serum | |
Sequential Triple-filtered, 200 nm | |
≤ 100 EU/mL | |
≤ 50 mg/dL |
Product Content Correction
Product Title
Cytiva HyClone™ Porcine Serum (New Zealand) > Irradiated; 1 L
Spot an opportunity for improvement?Share a Content Correction