Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
Brand:  Cytiva SH30333.02
 View more versions of this product
Product Code. 11572650
Description
Description
  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    cGMP
    Powder
    Non-sterile
    No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
    2° to 8°C, away from light
    PF-CHO MPS
    None
    10 L
    Product Title
    Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media > PF CHO MPS; Powder; 10L bottle