missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Synonyms: Hydrochloric acid fuming, Muriatic acid, Hydrogen chloride solution
- Molecular Formula: HCl
- Molar Mass: 36.46g/mol
- CAS: 7647-01-0
- EIN ECS No.: 231-595-7
- Residue After Evaporation: 0.005% max
Specifications
Specifications
pH | Passes test |
Appearance of Solution | Clear and colorless |
Melting Point | -28°C |
Boiling Point | ∼50°C |
Density | ∼1.19g/cm3 |
Heavy Metals (as Pb) | 0.0002% max |
Residue after Ignition | 0.003% max |
Vapor Pressure | 190hPa at 20°C |
Quantity | 1 L |
Physical Form | Solution |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Safety and Handling


Product Identifier
- Hydrochlorc acid, 37% extra pure, Pharmpur, Ph Eur, BP, NF
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Hydrogen chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrochloric Acid, 37%, Pharmpur™, Scharlab >
Spot an opportunity for improvement?Share a Content Correction