Learn More
(Hydroxy-2-naphthylmethyl)phosphonic acid, 98%, Thermo Scientific Chemicals
A selective insulin receptor tyrosine kinase inhibitor
132.66 EUR - 585.92 EUR
Chemical Identifiers
CAS | 132541-52-7 |
---|---|
Molecular Formula | C11H11O4P |
Molecular Weight (g/mol) | 238.179 |
MDL Number | MFCD00674174 |
InChI Key | AMJJLDJPDLKNJA-UHFFFAOYSA-N |
Synonym | hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010 |
PubChem CID | 129317 |
IUPAC Name | [hydroxy(naphthalen-2-yl)methyl]phosphonic acid |
SMILES | C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446069
|
Thermo Scientific Alfa Aesar
J63913.#0 |
5 mg |
132.66 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456069
|
Thermo Scientific Alfa Aesar
J63913.MB |
25 mg |
585.92 EUR
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A selective insulin receptor tyrosine kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
132541-52-7 | |
238.179 | |
AMJJLDJPDLKNJA-UHFFFAOYSA-N | |
129317 | |
C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O |
C11H11O4P | |
MFCD00674174 | |
hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010 | |
[hydroxy(naphthalen-2-yl)methyl]phosphonic acid |
Specifications
132541-52-7 | |
C11H11O4P | |
hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010 | |
C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O | |
238.179 | |
238.18 | |
Solid |
White | |
MFCD00674174 | |
AMJJLDJPDLKNJA-UHFFFAOYSA-N | |
[hydroxy(naphthalen-2-yl)methyl]phosphonic acid | |
129317 | |
98% | |
(Hydroxy-2-naphthylmethyl)phosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.