missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
- Hypericin is an anthraquinone derivative found in the yellow flower of Hypericum perforatum (also known as St. John's wort)
- This compound has the ability to inhibit protein kinase C, inducing photosynthesized apoptosis
Application
- It shows antibiotic and antiviral activities
- The antidepressant activity of this compound could be related to its ability to inhibit the neuronal uptake of serotonin, norepinephrine, dopamine, gamma-aminobutyric acid (GABA), and L-glutamate
- Since this compound is able to inhibit the assembly and shedding of virus particles in infected cells, hypericin may also prevent the replication of encapsulated viruses
- It has been described that this compound can also exhibit anti-HIV and anticancer activities
- Hypericin also displays an effective phototoxic activity and is able to trigger apoptotic pathways, resulting in the formation of reactive oxygen species

Specifications
Specifications
Quantity | 25mg |
Beilstein | 1917913 |
Sensitivity | Light sensitive |
Merck Index | 14,4863 |
Solubility Information | Soluble in ethanol,dimethyl sulfoxide,methanol,acetone,ethylmethylketone,pyridine and sodium hydroxide. Insoluble in water. |
Formula Weight | 504.44 |
Percent Purity | 98% |
Chemical Name or Material | Hypericin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S26K025 | Certificate of Analysis | 02/07/2024 | H26425.MC,H26425.MB |
10247112 | Certificate of Analysis | 22/06/2023 | H26425.MC,H26425.MB,H26425 |
10239884 | Certificate of Analysis | 12/04/2022 | H26425.MC,H26425.MB |
10187581 | Certificate of Analysis | 23/11/2021 | H26425.MC,H26425.MB |
A20X038 | Certificate of Analysis | 23/11/2021 | H26425.MC,H26425.MB |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hypericin, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction