Learn More
ICAM-2 Monoclonal Antibody (3C4), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A15451
201.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
ICAM-2 Monoclonal antibody specifically detects ICAM-2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
ICAM-2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P35330 | |
ICAM2 | |
Mouse CD102 from COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
3C4 | |
PE | |
ICAM2 | |
CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
Rat | |
purified | |
RUO | |
15896 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.