Learn More
IFI35 Monoclonal Antibody (OTI2G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503360

Description
Not yet known.Specifications
IFI35 | |
Monoclonal | |
0.41 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P80217 | |
IFI35 | |
Full length human recombit protein of human IFI35 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI2G5 | |
Unconjugated | |
IFI35 | |
IFP35 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3430 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.