missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech IHCeasy P53 Ready-To-Use IHC Kit
Ready-To-Use IHC Kit
Brand: Proteintech KHC0079-50T
This item is not returnable.
View return policy
Description
KHC0079 is a ready-to-use IHC kit for staining of P53. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.Specifications
Monoclonal | |
50 Tests | |
Mouse | |
|
Human | |
Immunohistochemistry (IHC) | |
All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech IHCeasy P53 Ready-To-Use IHC Kit > 50 Tests
Spot an opportunity for improvement?Share a Content Correction