missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-13 Porcine ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ESIL13X10
This item is not returnable.
View return policy
Description
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications
Q95J68 | |
16 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Il13 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
20.58 to 15,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
396721 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 20 μL|Serum, 20 μL|Supernatant, 50 μL | |
IL-13 | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ IL-13 Porcine ELISA Kit > 10 x 96 Tests
Spot an opportunity for improvement?Share a Content Correction