Learn More
Invitrogen™ Human IL-8 ELISA Development Kit (TMB), PeproTech®
ELISA
Brand:  Invitrogen™ 900-T18
Product Code. 17829165
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Interleukin 8 (IL-8, CXCL8) is a 72 amino acid pro-inflammatory factor which belongs to the CXC subfamily of chemokines, and are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C. IL-8 and ten other members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours, and IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P10145
HRP
Colorimetric Microplate Reader
CXCL8
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
ELISA
3576
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
RUO
IL-8
24 hrs 30 mins
Product Title
Invitrogen™ Human IL-8 ELISA Development Kit (TMB), PeproTech® > 10 x 96 Tests; HRP