missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL20RA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586848
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
Specifications
| IL20RA | |
| Polyclonal | |
| Unconjugated | |
| Il20ra | |
| class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 237313, 308716, 53832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PHB0, Q9UHF4 | |
| Il20ra | |
| A synthetic peptide corresponding to residues in Human IL20RA. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction