missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ile-Pro-Ile, 99.6%, For HPLC analysis, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
Specifications
Specifications
CAS | 90614-48-5 |
Molecular Formula | C17H31N3O4 |
Quantity | 25 mg |
Synonym | diprotin a, ile-pro-ile, isoleucylprolylisoleucine, n-1-l-isoleucyl-l-prolyl-l-isoleucine, l-isoleucine, n-1-l-isoleucyl-l-prolyl, l-isoleucyl-l-prolyl-l-isoleucine, spectrum2_001480, spectrum3_001838, isoleucyl-prolyl-isoleucine, d0n1qq |
InChI Key | JNTMAZFVYNDPLB-PEDHHIEDSA-N |
Molecular Weight (g/mol) | 341.45 |
PubChem CID | 94701 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ile-Pro-Ile, 99.6%, For HPLC analysis, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction